Assay Details
Target Gene Details
Entrez Gene ID: | 14634 |
Gene Name: | GLI-Kruppel family member GLI3 |
Gene Aliases: |
AI854843, AU023367, Bph, GLI3-190, GLI3FL, Pdn, Xt, add |
Location: |
Chr.13:15463235-15730026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gli3 | NM_008130.2 | NP_032156.2 | ||
| AK144851.1 | BAE26100.1 | |||
| AK158261.1 | BAE34433.1 | |||
| BC141135.1 | ||||
| BC145445.1 | ||||
| X95255.1 | CAA64543.1 |
More Information
Set Membership: |
|

Genomic Map