Assay Details
Target Gene Details
Entrez Gene ID: | 238871 |
Gene Name: | phosphodiesterase 4D, cAMP specific |
Gene Aliases: |
9630011N22Rik, Dpde3 |
Location: |
Chr.13:108654177-109955969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde4d | NM_011056.3 | NP_035186.1 | ||
| AF536978.1 | AAN10120.1 | |||
| BC132107.1 | ||||
| BC132109.1 |
More Information
Set Membership: |
|

Genomic Map