Assay Details
Target Gene Details
Entrez Gene ID: | 17121 |
Gene Name: | Max dimerization protein 3 |
Gene Aliases: |
4631412E13Rik, Mad3, bHLHc13 |
Location: |
Chr.13:55325168-55329770 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mxd3 | NM_016662.4 | NP_057871.2 | ||
| AK014516.1 | ||||
| AK086190.1 | ||||
| AK138742.1 | BAE23764.1 | |||
| BC051970.1 | AAH51970.1 | |||
| U32394.1 | AAB02794.1 |
More Information
Set Membership: |
|

Genomic Map