Assay Details
Target Gene Details
Entrez Gene ID: | 15357 |
Gene Name: | 3-hydroxy-3-methylglutaryl-Coenzyme A reductase |
Gene Aliases: |
HMG-CoAR, Red |
Location: |
Chr.13:96648962-96670936 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgcr | NM_008255.2 | NP_032281.2 | ||
| AK048784.1 | ||||
| AK159899.1 | ||||
| BC085083.1 | AAH85083.1 |
More Information
Set Membership: |
|

Genomic Map