Assay Details
Target Gene Details
Entrez Gene ID: | 15387 |
Gene Name: | heterogeneous nuclear ribonucleoprotein K |
Gene Aliases: |
Hnrpk, KBBP, NOVA |
Location: |
Chr.13:58391132-58403343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpk | NM_001301340.1 | NP_001288269.1 | ||
| NM_001301341.1 | NP_001288270.1 | |||
| NM_001301344.1 | NP_001288273.1 | |||
| NM_001301345.1 | NP_001288274.1 | |||
| NM_025279.3 | NP_079555.1 | |||
| AK011428.1 | BAB27614.1 | |||
| AK051313.1 | BAC34601.1 | |||
| AK078777.1 | BAC37389.1 | |||
| AK088462.1 | BAC40368.1 | |||
| AK128891.1 | ||||
| AK150848.1 | BAE29905.1 | |||
| AK151689.1 | BAE30613.1 | |||
| AK152927.1 | BAE31602.1 | |||
| AK160615.1 | BAE35915.1 | |||
| AK160887.1 | BAE36070.1 | |||
| AK166656.1 | BAE38921.1 | |||
| AK167600.1 | BAE39657.1 | |||
| AK169064.1 | BAE40851.1 | |||
| BC006694.1 | AAH06694.1 |
Target Gene Details
Entrez Gene ID: | 74386 |
Gene Name: | RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae) |
Gene Aliases: |
4932432N11Rik, C79893 |
Location: |
Chr.13:58402248-58411149 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rmi1 | NR_031761.1 | |||
| AK079457.1 | BAC37651.1 | |||
| AK139703.1 |
More Information
Set Membership: |
|

Genomic Map