Assay Details
Target Gene Details
Entrez Gene ID: | 328329 |
Gene Name: | microtubule associated serine/threonine kinase family member 4 |
Gene Aliases: |
4930420O11Rik, AI642422, mKIAA0303 |
Location: |
Chr.13:102732488-103335840 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mast4 | NM_175171.3 | NP_780380.2 | ||
| AK037351.1 | ||||
| AK090136.1 | ||||
| BC042511.1 | AAH42511.1 |
More Information
Set Membership: |
|

Genomic Map