Assay Details
Target Gene Details
Entrez Gene ID: | 18212 |
Gene Name: | neurotrophic tyrosine kinase, receptor, type 2 |
Gene Aliases: |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
Location: |
Chr.13:58806569-59133970 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntrk2 | NM_001025074.2 | 13 | 2181 | NP_001020245.1 |
| NM_001282961.1 | 13 | 2148 | NP_001269890.1 | |
| AK147391.1 | 13 | 1979 | BAE27880.1 | |
| AK160789.1 | 13 | 2149 | BAE36012.1 | |
| BG695150.1 | 2 | 153 |
More Information
Set Membership: |
|

Genomic Map