Assay Details
Target Gene Details
Entrez Gene ID: | 212880 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 46 |
Gene Aliases: |
2200005K02Rik, 8430438J23Rik, AI325430, AI957095, mKIAA0801 |
Location: |
Chr.13:55634969-55681267 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx46 | NM_001282055.1 | 1 | 114 | NP_001268984.1 |
AK008639.1 | 1 | 104 | BAB25800.3 | |
AK088950.1 | 1 | 86 | ||
AK129220.1 | 1 | 87 | BAC98030.2 | |
AK137835.1 | 1 | 104 | BAE23504.1 | |
BC019403.1 | 1 | 87 | ||
BC028552.1 | ||||
CJ170754.1 | 1 | 114 |
