Assay Details
Target Gene Details
Entrez Gene ID: | 212880 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 46 |
Gene Aliases: |
2200005K02Rik, 8430438J23Rik, AI325430, AI957095, mKIAA0801 |
Location: |
Chr.13:55634969-55681267 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx46 | NM_001282055.1 | 16 | 2225 | NP_001268984.1 |
| AK088950.1 | 15 | 3815 | ||
| AK129220.1 | 15 | 3816 | BAC98030.2 | |
| AK134073.1 | 4 | 1471 | BAE22002.1 | |
| BC026492.1 | 9 | 1170 | AAH26492.1 | |
| BC092240.1 | 15 | 2136 | AAH92240.1 |

Genomic Map