Assay Details
Target Gene Details
Entrez Gene ID: | 16398 |
Gene Name: | integrin alpha 2 |
Gene Aliases: |
CD49B, DX5 |
Location: |
Chr.13:114835912-114932041 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itga2 | NM_008396.2 | 8 | 994 | NP_032422.2 |
| AK036572.1 | 8 | 1054 | BAC29485.1 | |
| AK051050.1 | 8 | 1005 | ||
| AK156604.1 | 8 | 1036 | ||
| BC065139.1 | 8 | 994 | AAH65139.1 | |
| Z29987.1 | 8 | 998 | CAA82877.1 |
More Information
Set Membership: |
|

Genomic Map