Assay Details
Target Gene Details
Entrez Gene ID: | 212880 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 46 |
Gene Aliases: |
2200005K02Rik, 8430438J23Rik, AI325430, AI957095, mKIAA0801 |
Location: |
Chr.13:55634969-55681267 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx46 | NM_001282055.1 | 5 | 759 | NP_001268984.1 |
| AK088950.1 | 5 | 731 | ||
| AK129220.1 | 5 | 732 | BAC98030.2 | |
| BC092240.1 | 4 | 670 | AAH92240.1 |

Genomic Map