Assay Details
Target Gene Details
Entrez Gene ID: | 105348 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
2310001L02Rik, AW125446, D030064E01Rik, GP73, Golph2, PSEC0257 |
Location: |
Chr.13:59634996-59675784 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golm1 | NM_001035122.2 | 7 | 749 | NP_001030294.1 |
| NM_027307.4 | 8 | 885 | NP_081583.3 | |
| AK009072.1 | 6 | 556 | ||
| AK133865.1 | 7 | 751 | BAE21897.1 | |
| AK167646.1 | 7 | 726 | BAE39697.1 | |
| BC011152.1 | 7 | 710 | AAH11152.1 | |
| BC098486.1 | 8 | 881 | AAH98486.1 |

Genomic Map