Assay Details
Target Gene Details
Entrez Gene ID: | 238896 |
Gene Name: | cell division cycle 20B |
Gene Aliases: |
EG238896, EG622422 |
Location: |
Chr.13:113032954-113121019 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc20b |
Target Gene Details
Entrez Gene ID: | 14938 |
Gene Name: | granzyme A |
Gene Aliases: |
AW494114, Ctla-3, Ctla3, Hf, SE1, TSP-1, TSP1 |
Location: |
Chr.13:113093827-113100981 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gzma | NM_010370.2 | NP_034500.1 | ||
| AK146132.1 | BAE26923.1 | |||
| AK156728.1 | BAE33822.1 | |||
| BC061146.1 | AAH61146.1 | |||
| M13226.1 | AAA40134.1 | |||
| M26183.1 | AAA37735.1 | |||
| X14799.1 | CAA32905.1 |
More Information
Set Membership: |
|

Genomic Map