Assay Details
Target Gene Details
Entrez Gene ID: | 15487 |
Gene Name: | hydroxysteroid (17-beta) dehydrogenase 3 |
Gene Aliases: |
- |
Location: |
Chr.13:64058274-64089258 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd17b3 | NM_008291.3 | NP_032317.2 | ||
| BB621153.1 | ||||
| U66827.1 | AAB06793.1 |
More Information
Set Membership: |
|

Genomic Map