Assay Details
Target Gene Details
Entrez Gene ID: | 12919 |
Gene Name: | corticotropin releasing hormone binding protein |
Gene Aliases: |
CRH-BP |
Location: |
Chr.13:95431376-95444831 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crhbp | NM_198408.3 | NP_940800.1 | ||
| AK135067.1 | BAE22406.1 | |||
| AK139404.1 | ||||
| BC061247.1 | AAH61247.1 |

Genomic Map