Assay Details
Target Gene Details
Entrez Gene ID: | 78771 |
Gene Name: | multiple C2 domains, transmembrane 1 |
Gene Aliases: |
2810465F10Rik |
Location: |
Chr.13:76384961-77033284 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mctp1 | NM_030174.2 | 8 | 1015 | NP_084450.2 |
| AK047562.1 | 8 | 1016 | BAC33087.1 | |
| AK054478.1 | 4 | 388 | BAC35795.1 | |
| AK154322.1 | 7 | 830 | BAE32513.1 |

Genomic Map