Assay Details
Target Gene Details
Entrez Gene ID: | 17993 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 4 |
Gene Aliases: |
6720411N02Rik, C1-18k |
Location: |
Chr.13:114287795-114388094 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufs4 | NM_010887.2 | NP_035017.2 | ||
| AF124785.1 | AAD30474.1 | |||
| AK011124.1 | BAB27417.2 | |||
| AK020552.1 | BAB32133.3 | |||
| BC004618.1 | AAH04618.1 | |||
| CJ041886.1 | ||||
| CJ073528.1 |
More Information
Set Membership: |
|

Genomic Map