Assay Details
Target Gene Details
Entrez Gene ID: | 218624 |
Gene Name: | interleukin 31 receptor A |
Gene Aliases: |
CRL3, GLM-R, GPL, Glmr, NR10 |
Location: |
Chr.13:112521849-112591399 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il31ra | NM_139299.2 | 13 | 1721 | NP_647460.2 |
| AB083111.1 | 13 | 1720 | BAB88745.1 | |
| AF486621.1 | 10 | 1300 | AAM27959.1 | |
| AK030512.1 | 2 | 290 | BAC26998.1 | |
| BC066164.1 | 11 | 1442 | AAH66164.1 |
More Information
Set Membership: |
|

Genomic Map