Assay Details
Target Gene Details
Entrez Gene ID: | 218624 |
Gene Name: | interleukin 31 receptor A |
Gene Aliases: |
CRL3, GLM-R, GPL, Glmr, NR10 |
Location: |
Chr.13:112521849-112591399 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il31ra | NM_139299.2 | NP_647460.2 | ||
| AB083111.1 | BAB88745.1 | |||
| AF486621.1 | AAM27959.1 | |||
| BC066164.1 | AAH66164.1 |
More Information
Set Membership: |
|

Genomic Map