Assay Details
Target Gene Details
Entrez Gene ID: | 19720 |
Gene Name: | tripartite motif-containing 27 |
Gene Aliases: |
AW538890, Gm19403, Rfp |
Location: |
Chr.13:21179440-21194723 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim27 | NM_009054.3 | NP_033080.2 | ||
| AK031744.1 | ||||
| AK088180.1 | BAC40192.1 | |||
| BC003219.1 | AAH03219.1 | |||
| BC069924.1 | AAH69924.1 | |||
| CA549428.1 | ||||
| X75343.1 | CAA53092.1 |
More Information
Set Membership: |
|

Genomic Map