Assay Details
Target Gene Details
Entrez Gene ID: | 20726 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 9d |
Gene Aliases: |
AT2, Spi9, ovalbumin |
Location: |
Chr.13:33192959-33203980 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb9d | NM_011460.2 | NP_035590.1 | ||
| AK028382.1 | BAC25920.1 | |||
| BC139261.1 | ||||
| BC139263.1 |
More Information
Set Membership: |
|

Genomic Map