Assay Details
Target Gene Details
Entrez Gene ID: | 74188 |
Gene Name: | prolactin family 8, subfamily a, member 81 |
Gene Aliases: |
1600032B14Rik, AV036230, PLP-Cgamma, Prlpc3 |
Location: |
Chr.13:27507032-27513228 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl8a8 | NM_023741.2 | NP_076230.1 | ||
| AF230921.1 | AAN76491.1 | |||
| AF230922.1 | AAN76492.1 | |||
| AF230923.1 | AAN76493.1 | |||
| AF466150.1 | AAL73987.1 | |||
| AK005568.1 | BAB24128.1 | |||
| AK167369.1 | BAE39465.1 | |||
| BC100326.1 | AAI00327.1 |
More Information
Set Membership: |
|

Genomic Map