Assay Details
Target Gene Details
Entrez Gene ID: | 18708 |
Gene Name: | phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha) |
Gene Aliases: |
PI3K, p50alpha, p55alpha, p85alpha |
Location: |
Chr.13:101680761-101768217 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3r1 | NM_001024955.2 | 4 | 738 | NP_001020126.1 |
| NM_001077495.2 | 10 | 1827 | NP_001070963.1 | |
| AK046259.1 | 4 | 740 | ||
| BC026146.1 | 10 | 1808 | AAH26146.1 | |
| BC051106.1 | 4 | 438 | AAH51106.1 | |
| CB519962.1 | 4 | 370 | ||
| M60651.1 | 10 | 1762 | AAA39886.1 | |
| U50413.1 | 10 | 1780 | AAC52847.1 | |
| U50414.1 | AAC52848.1 |
More Information
Set Membership: |
|

Genomic Map