Assay Details
Target Gene Details
Entrez Gene ID: | 18708 |
Gene Name: | phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha) |
Gene Aliases: |
PI3K, p50alpha, p55alpha, p85alpha |
Location: |
Chr.13:101680761-101768217 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3r1 | NM_001024955.2 | NP_001020126.1 | ||
| NM_001077495.2 | NP_001070963.1 | |||
| AK046259.1 | ||||
| BC026146.1 | AAH26146.1 | |||
| BC051106.1 | AAH51106.1 | |||
| CB519962.1 | ||||
| M60651.1 | AAA39886.1 | |||
| U50413.1 | AAC52847.1 |
More Information
Set Membership: |
|

Genomic Map