Assay Details
Target Gene Details
Entrez Gene ID: | 432763 |
Gene Name: | proline rich 7 (synaptic) |
Gene Aliases: |
XM_484263 |
Location: |
Chr.13:55464267-55473155 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prr7 | NM_001030296.4 | 1 | 97 | NP_001025467.1 |
| AK133129.1 | BAE21522.1 | |||
| CX241234.1 | 1 | 97 |

Genomic Map