Assay Details
Target Gene Details
Entrez Gene ID: | 20418 |
Gene Name: | src homology 2 domain-containing transforming protein C3 |
Gene Aliases: |
N-Shc, Rai, ShcC |
Location: |
Chr.13:51431041-51567084 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shc3 | NM_009167.3 | NP_033193.2 | ||
| BC103611.1 | AAI03612.1 | |||
| BC103612.1 | AAI03613.1 | |||
| BC105644.1 | ||||
| BC105645.1 | ||||
| U46854.1 | AAC52508.1 |
More Information
Set Membership: |
|

Genomic Map