Assay Details
Target Gene Details
Entrez Gene ID: | 218460 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
B830029I03Rik, MSTP048 |
Location: |
Chr.13:94976213-95023316 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr41 | NM_172590.3 | 12 | 1241 | NP_766178.2 |
| AK046853.1 | 12 | 1210 | BAC32897.1 | |
| AK149993.1 | 12 | 1207 | BAE29221.1 | |
| AK194357.1 | 3 | 281 |

Genomic Map