Assay Details
Target Gene Details
Entrez Gene ID: | 218460 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
B830029I03Rik, MSTP048 |
Location: |
Chr.13:94976213-95023316 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr41 | NM_172590.3 | 2 | 307 | NP_766178.2 |
| AK046853.1 | 2 | 276 | BAC32897.1 | |
| AK082348.1 | 2 | 308 | BAC38474.1 | |
| AK149993.1 | 2 | 273 | BAE29221.1 | |
| AK194741.1 | ||||
| BB650073.1 | 2 | 303 | ||
| BC138095.1 | 2 | 213 | ||
| BC145144.1 | 2 | 213 |

Genomic Map