Assay Details
Target Gene Details
Entrez Gene ID: | 218460 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
B830029I03Rik, MSTP048 |
Location: |
Chr.13:94976213-95023316 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr41 | NM_172590.3 | 4 | 479 | NP_766178.2 |
| AK046853.1 | 4 | 448 | BAC32897.1 | |
| AK082348.1 | 4 | 480 | BAC38474.1 | |
| AK149993.1 | 4 | 445 | BAE29221.1 | |
| AK194741.1 | 3 | 146 | ||
| BB650073.1 | 4 | 475 | ||
| BC138095.1 | 4 | 385 | ||
| BC145144.1 | 4 | 385 |

Genomic Map