Assay Details
Target Gene Details
Entrez Gene ID: | 12294 |
Gene Name: | calcium channel, voltage-dependent, alpha2/delta subunit 3 |
Gene Aliases: |
Cacnad3 |
Location: |
Chr.14:28904943-29721864 on Build GRCm38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna2d3 | NM_009785.1 | NP_033915.1 | ||
| AJ010949.1 | CAA09423.1 |
More Information
Set Membership: |
|

Genomic Map