Assay Details
Target Gene Details
Entrez Gene ID: | 19166 |
Gene Name: | proteasome (prosome, macropain) subunit, alpha type 2 |
Gene Aliases: |
Lmpc3 |
Location: |
Chr.13:14613242-14625673 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psma2 | NM_008944.2 | NP_032970.2 | ||
| AF099733.1 | AAD50623.1 | |||
| AK012119.1 | BAB28045.1 | |||
| AK012990.1 | BAB28582.1 | |||
| AK136115.1 | BAE22828.1 | |||
| AK151883.1 | BAE30769.1 | |||
| AK152862.1 | BAE31553.1 | |||
| AK154011.1 | BAE32314.1 | |||
| AK167752.1 | BAE39787.1 | |||
| AK209655.1 | ||||
| BC026768.1 | AAH26768.1 | |||
| BU755659.1 | ||||
| CJ052982.1 | ||||
| X70303.1 | CAA49782.1 |
More Information
Set Membership: |
|

Genomic Map