Assay Details
Target Gene Details
Entrez Gene ID: | 72198 |
Gene Name: | superkiller viralicidic activity 2-like 2 (S. cerevisiae) |
Gene Aliases: |
2610528A15Rik, mKIAA0052 |
Location: |
Chr.13:112867780-112927380 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skiv2l2 | NM_028151.2 | 15 | 1612 | NP_082427.1 |
| AK012158.1 | 15 | 1612 | BAB28066.1 | |
| AK078091.1 | 15 | 1631 | BAC37122.1 | |
| AK088426.1 | 14 | 1449 | BAC40348.1 | |
| AK129045.1 | 8 | 767 | BAC97855.2 | |
| BC014810.1 | 7 | 646 | AAH14810.1 | |
| BC029230.1 | 15 | 1602 | AAH29230.1 |
More Information
Set Membership: |
|

Genomic Map