Assay Details
Target Gene Details
Entrez Gene ID: | 239096 |
Gene Name: | cadherin-like 24 |
Gene Aliases: |
1700040A22Rik, ENSMUSG00000022188 |
Location: |
Chr.14:54631992-54641364 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh24 | NM_199470.2 | NP_955764.1 |
More Information
Set Membership: |
|

Genomic Map