Assay Details
Target Gene Details
Entrez Gene ID: | 214189 |
Gene Name: | secretagogin, EF-hand calcium binding protein |
Gene Aliases: |
- |
Location: |
Chr.13:23953456-23991337 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scgn | NM_145399.1 | NP_663374.1 | ||
| AK148468.1 | ||||
| BC016093.1 | AAH16093.1 |
More Information
Set Membership: |
|

Genomic Map