Assay Details
Target Gene Details
Entrez Gene ID: | 72198 |
Gene Name: | superkiller viralicidic activity 2-like 2 (S. cerevisiae) |
Gene Aliases: |
2610528A15Rik, mKIAA0052 |
Location: |
Chr.13:112867780-112927380 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skiv2l2 | NM_028151.2 | NP_082427.1 | ||
| AK012158.1 | BAB28066.1 | |||
| AK088426.1 | BAC40348.1 | |||
| AK129045.1 | BAC97855.2 | |||
| BC014810.1 | AAH14810.1 | |||
| BC029230.1 | AAH29230.1 |
More Information
Set Membership: |
|

Genomic Map