Assay Details
Target Gene Details
Entrez Gene ID: | 72391 |
Gene Name: | cyclin-dependent kinase inhibitor 3 |
Gene Aliases: |
2410006H10Rik, KAP |
Location: |
Chr.14:46760464-46771529 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn3 | NM_028222.1 | NP_082498.1 | ||
| AK010426.1 | BAB26929.1 | |||
| AK033341.1 | BAC28238.1 | |||
| AK146312.1 | BAE27067.1 | |||
| BC049694.1 | AAH49694.1 | |||
| BY084534.1 |
More Information
Set Membership: |
|

Genomic Map