Assay Details
Target Gene Details
Entrez Gene ID: | 72198 |
Gene Name: | superkiller viralicidic activity 2-like 2 (S. cerevisiae) |
Gene Aliases: |
2610528A15Rik, mKIAA0052 |
Location: |
Chr.13:112867780-112927380 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skiv2l2 | NM_028151.2 | 5 | 485 | NP_082427.1 |
| AK012158.1 | 5 | 485 | BAB28066.1 | |
| AK077924.1 | 5 | 477 | BAC37067.1 | |
| AK078091.1 | 5 | 504 | BAC37122.1 | |
| AK088426.1 | 4 | 322 | BAC40348.1 | |
| BC029230.1 | 5 | 475 | AAH29230.1 |
More Information
Set Membership: |
|

Genomic Map