Assay Details
Target Gene Details
Entrez Gene ID: | 12143 |
Gene Name: | B lymphoid kinase |
Gene Aliases: |
- |
Location: |
Chr.14:63372831-63417187 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blk | NM_007549.2 | NP_031575.2 | ||
| AK089888.1 | BAC40986.1 | |||
| AK171640.1 | BAE42580.1 | |||
| BC030668.1 | AAH30668.1 | |||
| CT010240.1 | CAJ18448.1 | |||
| M30903.1 | AAA40453.1 |

Genomic Map