Assay Details
Target Gene Details
Entrez Gene ID: | 76633 |
Gene Name: | RIKEN cDNA 1700112E06 gene |
Gene Aliases: |
2700009F18Rik |
Location: |
Chr.14:22019503-23056088 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700112E06Rik | NM_028275.1 | NP_082551.1 | ||
| AK007175.1 | BAB24884.1 | |||
| AK153491.1 | BAE32039.1 |
Target Gene Details
Entrez Gene ID: | 791307 |
Gene Name: | predicted gene 10248 |
Gene Aliases: |
ENSMUSG00000068680 |
Location: |
Chr.14:23038194-23094571 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10248 | NR_033550.1 | |||
| AY512910.1 | AAR87781.1 |
More Information
Set Membership: |
|

Genomic Map