Assay Details
Target Gene Details
Entrez Gene ID: | 66494 |
Gene Name: | PRELI domain containing 1 |
Gene Aliases: |
2610524G07Rik, Preli |
Location: |
Chr.13:55322055-55325272 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prelid1 | NM_025596.5 | NP_079872.4 | ||
| AK002846.1 | ||||
| AK003629.1 | BAB22898.2 | |||
| AK012144.1 | ||||
| AK146857.1 | BAE27486.1 | |||
| AK150465.1 | BAE29582.1 | |||
| AK151321.1 | BAE30302.1 | |||
| AK153455.1 | ||||
| AK155042.1 | BAE33009.1 | |||
| AK196389.1 | ||||
| BC024813.1 | ||||
| BC025859.1 | AAH25859.1 | |||
| BC098241.1 | AAH98241.1 |
More Information
Set Membership: |
|

Genomic Map