Assay Details
Target Gene Details
Entrez Gene ID: | 26564 |
Gene Name: | receptor tyrosine kinase-like orphan receptor 2 |
Gene Aliases: |
Ntrkr2, mRor2 |
Location: |
Chr.13:53109317-53286109 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ror2 | NM_013846.3 | NP_038874.3 | ||
| AB010384.1 | BAA75481.1 | |||
| AK031112.1 | BAC27258.1 | |||
| AK031789.1 | ||||
| AK083538.1 | ||||
| AK084752.1 | BAC39273.1 | |||
| AK157705.1 | BAE34159.1 | |||
| BC030848.1 | AAH30848.1 | |||
| BC070436.1 | AAH70436.1 |
More Information
Set Membership: |
|

Genomic Map