Assay Details
Target Gene Details
Entrez Gene ID: | 105372 |
Gene Name: | UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast) |
Gene Aliases: |
AW544865 |
Location: |
Chr.13:98246845-98262992 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Utp15 | NM_178918.3 | 11 | 1410 | NP_849249.1 |
| AK049186.1 | 11 | 1410 | BAC33596.1 | |
| AK162794.1 | 11 | 1414 | BAE37061.1 | |
| AK165546.1 | 11 | 1405 | BAE38247.1 | |
| AY151261.1 | 11 | 1351 | AAN71642.1 | |
| BC034372.1 | 7 | 767 | AAH34372.1 | |
| BC065057.1 | 11 | 1410 | AAH65057.1 |
More Information
Set Membership: |
|

Genomic Map