Assay Details
Target Gene Details
Entrez Gene ID: | 78925 |
Gene Name: | steroid 5 alpha-reductase 1 |
Gene Aliases: |
0610031P22Rik, 4930435F02Rik, S5AR 1, Srd5a-1 |
Location: |
Chr.13:69573449-69611900 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srd5a1 | NM_175283.3 | 3 | 712 | NP_780492.2 |
| AK082819.1 | 3 | 604 | BAC38634.1 | |
| BC079863.1 | 3 | 690 | AAH79863.1 | |
| BC115468.1 | 3 | 569 | ||
| BC115469.1 | 3 | 569 |
More Information
Set Membership: |
|

Genomic Map