Assay Details
Target Gene Details
Entrez Gene ID: | 105513 |
Gene Name: | charged multivesicular body protein 7 |
Gene Aliases: |
4930596K11Rik, 6330407G04Rik, AI450338, AW550775 |
Location: |
Chr.14:69716979-69732570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp7 | NM_134078.4 | NP_598839.2 | ||
| BB621791.1 | ||||
| BC024115.1 | AAH24115.1 | |||
| BC033365.1 | AAH33365.1 |
More Information
Set Membership: |
|

Genomic Map