Assay Details
Target Gene Details
Entrez Gene ID: | 18776 |
Gene Name: | prolactin family 3, subfamily b, member 1 |
Gene Aliases: |
Csh2, PL, Pl-2, Pl2, mPL-II, mplII |
Location: |
Chr.13:27242430-27249740 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl3b1 | NM_008865.3 | NP_032891.1 | ||
| AK005542.1 | BAB24111.1 | |||
| AK005543.1 | BAB24112.1 | |||
| AK131667.1 | BAE20754.1 | |||
| AK167483.1 | BAE39564.1 | |||
| BC099386.1 | AAH99386.1 | |||
| M14647.1 | AAA39947.1 |
More Information
Set Membership: |
|

Genomic Map