Assay Details
Target Gene Details
Entrez Gene ID: | 13003 |
Gene Name: | versican |
Gene Aliases: |
5430420N07Rik, 9430051N09, Cspg2, DPEAAE, NG2, PG-M, PG-M(V0), PG-M(V1), hdf |
Location: |
Chr.13:89655309-89742694 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vcan | NM_001081249.1 | 10 | 9752 | NP_001074718.1 |
| NM_001134474.1 | 9 | 4535 | NP_001127946.1 | |
| NM_001134475.1 | 8 | 1655 | NP_001127947.1 | |
| NM_019389.2 | 9 | 6872 | NP_062262.2 | |
| AK045314.1 | 3 | 3207 | ||
| D16263.1 | 8 | 6444 | BAA03796.1 | |
| D28599.1 | 10 | 9496 | ||
| D32040.1 | 8 | 1387 | BAA06802.1 |
More Information
Set Membership: |
|

Genomic Map