Assay Details
Target Gene Details
Entrez Gene ID: | 72400 |
Gene Name: | PIN2/TERF1 interacting, telomerase inhibitor 1 |
Gene Aliases: |
2210403I16Rik, 2610028A01Rik, 67-11-3, AU024023, LPTS, LPTS1 |
Location: |
Chr.14:63860301-63919862 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pinx1 | NM_028228.3 | NP_082504.1 | ||
| AF321817.1 | AAL37221.1 | |||
| AF421879.1 | AAL32445.1 | |||
| AJ344106.1 | CAC51439.1 | |||
| AK012057.1 | BAE43228.1 | |||
| AK075840.1 | BAC35998.1 | |||
| BC115636.1 |
More Information
Set Membership: |
|

Genomic Map