Assay Details
Target Gene Details
Entrez Gene ID: | 12224 |
Gene Name: | Kruppel-like factor 5 |
Gene Aliases: |
4930520J07Rik, Bteb2, CKLF, IKLF |
Location: |
Chr.14:99298691-99313412 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf5 | NM_009769.4 | NP_033899.2 | ||
| AB025099.3 | BAA92284.3 | |||
| AF079852.1 | AAD17696.1 | |||
| AK015853.1 | 1 | 976 | ||
| AK051409.1 | ||||
| AK087066.1 | ||||
| BC006646.1 | AAH06646.1 | |||
| BC012958.1 | AAH12958.1 |
More Information
Set Membership: |
|

Genomic Map