Assay Details
Target Gene Details
Entrez Gene ID: | 218977 |
Gene Name: | discs, large (Drosophila) homolog-associated protein 5 |
Gene Aliases: |
C77459, C86398, Dap-5, Dlg7, Hurp, mKIAA0008 |
Location: |
Chr.14:47387779-47418407 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlgap5 | NM_144553.2 | NP_653136.2 | ||
| AB076696.1 | BAB97377.1 | |||
| AK036384.1 | BAC29405.1 | |||
| AK048470.1 | ||||
| AK085882.1 | BAC39560.1 | |||
| AK129032.1 | BAC97842.1 | |||
| AK135820.1 | BAE22675.1 | |||
| AK147955.1 | BAE28249.1 | |||
| BC043924.1 | AAH43924.1 | |||
| BC058087.1 | AAH58087.1 |
More Information
Set Membership: |
|

Genomic Map