Assay Details
Target Gene Details
Entrez Gene ID: | 268709 |
Gene Name: | family with sequence similarity 107, member A |
Gene Aliases: |
DRR1 |
Location: |
Chr.14:8296270-8318018 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam107a | NM_183187.3 | NP_899010.1 | ||
| AK044219.1 | BAC31824.1 | |||
| AK165722.1 | ||||
| BC055107.1 | AAH55107.1 |

Genomic Map